siRNA Details
| Gene Symbol: RELL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085399.1 | NP_001078868.1 | 4 | 489 | ||
| NM_001085400.1 | NP_001078869.1 | 4 | 489 | |||
| XM_017008590.1 | XP_016864079.1 | 2 | 157 | |||
| GenBank mRNA | AK075468.1 | - | 4 | 488 | ||
| BC039540.1 | - | 4 | 487 | 5748847 | ||
| DA400382.1 | - | 4 | 489 | |||

RELL1
Genomic Map