siRNA Details
| Gene Symbol: PSAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085382.1 | NP_001078851.1 | 1 | 1418 | ||
| GenBank mRNA | AK097698.1 | - | 1 | 701 | ||
| BC034552.1 | - | 1 | 1333 | |||
| BC068579.1 | - | 1 | 1392 | |||
| DQ991252.1 | - | 1 | 1418 | |||

PSAPL1
Genomic Map