siRNA Details
| Gene Symbol: Abca4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007378.1 | NP_031404.1 | 8 | 1169 | ||
| GenBank mRNA | AF000149.1 | - | 8 | 1169 | ||
| AK053043.1 | - | 3 | 450 | |||
| AK084860.1 | - | 9 | 1397 | |||
| BC057853.1 | - | 8 | 1169 | 6492139 | ||

Abca4
Genomic Map