siRNA Details
| Gene Symbol: Abcg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009593.2 | NP_033723.1 | 14 | 1824 | ||
| GenBank mRNA | AF323659.1 | - | 14 | 1820 | ||
| BC119471.2 | - | 14 | 1772 | 40048166 | ||
| CO431055.1 | - | 2 | 69 | |||
| CV558133.1 | - | 6 | 635 | |||
| U34920.1 | - | 14 | 1782 | |||
| Z48745.1 | - | 14 | 1811 | |||

Abcg1
Genomic Map