siRNA Details
| Gene Symbol: Scgb1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009596.1 | NP_033726.1 | 2 | 153 | ||
| GenBank mRNA | BC016132.1 | - | 2 | 153 | 4922791 | |
| BC132436.1 | - | 2 | 237 | 40130713 | ||
| GU269231.1 | - | 2 | 152 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scgb1b27| Gene Symbol: Scgb1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009596.1 | NP_033726.1 | 2 | 153 | ||
| GenBank mRNA | BC016132.1 | - | 2 | 153 | 4922791 | |
| BC132436.1 | - | 2 | 237 | 40130713 | ||
| GU269231.1 | - | 2 | 152 | |||