siRNA Details
| Gene Symbol: Scgb1b27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009596.1 | NP_033726.1 | 2 | 215 | ||
| GenBank mRNA | AF039064.1 | - | 1 | 148 | ||
| BC016132.1 | - | 2 | 215 | 4922791 | ||
| BC132436.1 | - | 2 | 299 | 40130713 | ||
| GU269231.1 | - | 2 | 214 | |||

Scgb1b27
Genomic Map