siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 8 | 843 | ||
| GenBank mRNA | AK131793.1 | - | 8 | 645 | ||
| AK146444.1 | - | 8 | 648 | |||
| U07159.1 | - | 8 | 632 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acadm| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 8 | 843 | ||
| GenBank mRNA | AK131793.1 | - | 8 | 645 | ||
| AK146444.1 | - | 8 | 648 | |||
| U07159.1 | - | 8 | 632 | |||