siRNA Details
| Gene Symbol: Acadm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007382.5 | NP_031408.1 | 3, 4 | 445 | ||
| GenBank mRNA | AK131793.1 | - | 3, 4 | 247 | ||
| AK146444.1 | - | 3, 4 | 250 | |||
| BC013498.1 | - | 3, 4 | 230 | 4222450 | ||
| CX232934.1 | - | 3, 4 | 436 | |||
| U07159.1 | - | 3, 4 | 234 | |||

Acadm
Genomic Map