siRNA Details
| Gene Symbol: Acadvl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_017366.3 | NP_059062.1 | 14, 15 | 1537 | ||
| GenBank mRNA | AF017176.1 | - | 11, 12 | 1185 | ||
| AK076037.1 | - | 14, 15 | 1498 | |||
| AK146334.1 | - | 9, 10 | 1010 | |||
| AK180918.1 | - | 2, 3 | 157 | |||
| U41497.1 | - | 5, 6 | 419 | |||
| Z71189.1 | - | 14, 15 | 1458 | |||

Acadvl
Genomic Map