siRNA Details
| Gene Symbol: Acads | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007383.3 | NP_031409.2 | 5 | 629 | ||
| GenBank mRNA | AK155361.1 | - | 5 | 608 | ||
| AK169428.1 | - | 5 | 566 | |||
| BC016259.1 | - | 5 | 596 | 4913813 | ||
| BC055063.1 | - | 5 | 570 | |||
| L11163.1 | - | 5 | 553 | |||

Acads
Genomic Map