siRNA Details
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009603.1 | NP_033733.1 | 5 | 404 | ||
GenBank mRNA | X55718.1 | - | 5 | 404 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Chrne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009603.1 | NP_033733.1 | 5 | 404 | ||
GenBank mRNA | X55718.1 | - | 5 | 404 |