siRNA Details
| Gene Symbol: GRM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278064.1 | NP_001264993.1 | 7 | 2141 | ||
| NM_001278065.1 | NP_001264994.1 | 8 | 2188 | |||
| NM_001278066.1 | NP_001264995.1 | 7 | 2141 | |||
| NM_001278067.1 | NP_001264996.1 | 7 | 2141 | |||
| XM_011535782.1 | XP_011534084.1 | 8 | 2291 | |||
| XM_017010783.1 | XP_016866272.1 | 8 | 2300 | |||
| XM_017010784.1 | XP_016866273.1 | 8 | 2336 | |||
| XM_017010785.1 | XP_016866274.1 | 8 | 2296 | |||
| XM_017010786.1 | XP_016866275.1 | 8 | 2296 | |||
| XM_017010787.1 | XP_016866276.1 | 8 | 2332 | |||
| XM_017010788.1 | XP_016866277.1 | 6 | 1108 | |||
| GenBank mRNA | AB208837.1 | - | 7 | 1808 | ||
| BC136280.1 | - | 7 | 1853 | 9020267 | ||
| BC143777.1 | - | 7 | 1853 | |||
| BC143779.1 | - | 7 | 1851 | |||
| L76627.1 | - | 7 | 1821 | |||
| L76631.1 | - | 8 | 2191 | |||
| U31215.1 | - | 7 | 2056 | |||
| U31216.1 | - | 7 | 1821 | |||
More Information
Set Membership: |
|

GRM1
Genomic Map