siRNA Details
| Gene Symbol: Ada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001272052.1 | NP_001258981.1 | 4 | 482 | ||
| NM_007398.4 | NP_031424.1 | 4 | 482 | |||
| GenBank mRNA | AF483480.1 | - | 4 | 334 | ||
| AF483481.1 | - | 4 | 334 | |||
| AK075899.1 | - | 4 | 421 | |||
| AK167489.1 | - | 4 | 421 | |||
| BC002075.1 | - | 4 | 432 | 3484465 | ||
| CT010224.1 | - | 4 | 334 | |||
| CT010255.1 | - | 4 | 334 | |||
| M10319.1 | - | 4 | 404 | |||

Ada
Genomic Map