siRNA Details
| Gene Symbol: Adam10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007399.3 | NP_031425.2 | 9 | 1301 | ||
| GenBank mRNA | AF011379.1 | - | 9 | 1122 | ||
| AK036599.1 | - | 3 | 345 | |||
| BC057346.1 | - | 9 | 1262 | |||
| BC066207.1 | - | 9 | 1306 | |||

Adam10
Genomic Map