siRNA Details
| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007402.2 | NP_031428.2 | 16 | 1788 | ||
| GenBank mRNA | AF013107.1 | - | 16 | 1774 | ||
| AK136692.1 | - | 16 | 1787 | |||
| AK164965.1 | - | 1 | 237 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Adam7| Gene Symbol: Adam7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007402.2 | NP_031428.2 | 16 | 1788 | ||
| GenBank mRNA | AF013107.1 | - | 16 | 1774 | ||
| AK136692.1 | - | 16 | 1787 | |||
| AK164965.1 | - | 1 | 237 | |||