siRNA Details
| Gene Symbol: Add2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271857.1 | NP_001258786.1 | 16 | 2451 | ||
| NM_001271858.1 | NP_001258787.1 | 16 | 2448 | |||
| NM_001271859.1 | NP_001258788.1 | 16 | 2249 | |||
| NM_013458.5 | NP_038486.2 | 16 | 2252 | |||
| GenBank mRNA | AF100422.1 | - | 15 | 2127 | ||
| AF189769.1 | - | 14 | 2049 | |||
| AF189770.1 | - | 9 | 1305 | |||
| AK014496.1 | - | 16 | 2217 | |||
| AK029488.1 | - | 16 | 2402 | |||
| AK147544.1 | - | 16 | 2329 | |||
| AK156954.1 | - | 16 | 2241 | |||

Add2
Genomic Map