siRNA Details
| Gene Symbol: Adh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007409.2 | NP_031435.1 | 6 | 811 | ||
| GenBank mRNA | AK146094.1 | - | 6 | 812 | ||
| BC013477.1 | - | 6 | 811 | 4238555 | ||
| BC054467.1 | - | 6 | 791 | 6530906 | ||
| M22611.1 | - | 1 | 70 | |||

Adh1
Genomic Map