siRNA Details
| Gene Symbol: Adh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001288578.1 | NP_001275507.1 | 6 | 741 | ||
| NM_007410.3 | NP_031436.2 | 6 | 759 | |||
| GenBank mRNA | AK002790.1 | - | 6 | 681 | ||
| AK076507.1 | - | 6 | 688 | |||
| AK146949.1 | - | 6 | 703 | |||
| AK159803.1 | - | 6 | 739 | |||
| AK214149.1 | - | 2 | 217 | |||
| BC062879.1 | - | 6 | 685 | 5691327 | ||
| BC090978.1 | - | 6 | 682 | 30313517 | ||
| M84147.1 | - | 6 | 709 | |||

Adh5
Genomic Map