siRNA Details
| Gene Symbol: Gpr182 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007412.2 | NP_031438.2 | 2 | 319 | ||
| GenBank mRNA | AK031480.1 | - | 254 | |||
| BC125279.1 | - | 2 | 190 | |||
| BY121440.1 | - | 2 | 320 | |||
| D17292.1 | - | 2 | 214 | |||
More Information
Set Membership: |
|

Gpr182
Genomic Map