siRNA Details
| Gene Symbol: Adra1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001284380.1 | NP_001271309.1 | 3 | 1063 | ||
| NM_001284381.1 | NP_001271310.1 | 4 | 1213 | |||
| NM_007416.4 | NP_031442.2 | 1 | 1059 | |||
| GenBank mRNA | AK004891.1 | - | 4 | 1170 | ||
| AK043877.1 | - | 1 | 904 | |||
| AK139448.1 | - | 1 | 1062 | |||
| BC018330.1 | - | 3 | 1036 | |||
| BC037002.1 | - | 3 | 1037 | |||
| BC138653.1 | - | 1 | 777 | 8861673 | ||
| BC138654.1 | - | 1 | 777 | 8861674 | ||
| Y12738.1 | - | 1 | 1378 | |||
More Information
Set Membership: |
|

Adra1b
Genomic Map