siRNA Details
| Gene Symbol: Adra2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009633.3 | NP_033763.2 | 1 | 804 | ||
| GenBank mRNA | AF332049.1 | - | 1 | 510 | ||
| AF332050.1 | - | 1 | 510 | |||
| BC066862.1 | - | 1 | 804 | |||
More Information
Set Membership: |
|

Adra2b
Genomic Map