siRNA Details
Gene Symbol: Adra2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007418.3 | NP_031444.2 | 1 | 1494 |

More Information
Set Membership: |
![]() |
Panther Classification:
Gene Ontology Categories:
