siRNA Details
| Gene Symbol: Agtr1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175086.3 | NP_780295.2 | 2 | 853 | ||
| GenBank mRNA | AK087228.1 | - | 2 | 853 | ||
| BC109152.1 | - | 2 | 800 | |||
| BC109153.1 | - | 2 | 800 | 40046982 | ||
More Information
Set Membership: |
|

Agtr1b
Genomic Map