siRNA Details
| Gene Symbol: Ak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198790.1 | NP_001185719.1 | 4 | 234 | ||
| NM_001198791.1 | NP_001185720.1 | 4 | 216 | |||
| NM_001198792.1 | NP_001185721.1 | 3 | 175 | |||
| NM_021515.3 | NP_067490.1 | 3 | 193 | |||
| GenBank mRNA | AJ010108.1 | - | 3 | 99 | ||
| AJ010109.1 | - | 3 | 189 | |||
| AK046613.1 | - | 4 | 235 | |||
| AK089270.1 | - | 4 | 217 | |||
| BC014802.1 | - | 4 | 160 | 4505142 | ||
| BC054366.1 | - | 3 | 182 | 5346232 | ||
| BY130582.1 | - | 4 | 217 | |||
| BY333336.1 | - | 3 | 175 | |||
| CF950379.1 | - | 3 | 193 | |||
| DQ486026.1 | - | 4 | 247 | |||

Ak1
Genomic Map