siRNA Details
| Gene Symbol: Ak4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177602.1 | NP_001171073.1 | 5 | 904 | ||
| NM_001177604.1 | NP_001171075.1 | 5 | 717 | |||
| NM_001177605.1 | NP_001171076.1 | 5 | 695 | |||
| NM_009647.5 | NP_033777.1 | 4 | 662 | |||
| GenBank mRNA | AB020239.1 | - | 4 | 626 | ||
| AK134600.1 | - | 5 | 907 | |||
| AK144439.1 | - | 4 | 632 | |||
| AK154374.1 | - | 5 | 721 | |||
| AK166975.1 | - | 4 | 584 | |||
| BC086663.1 | - | 4 | 610 | 30649610 | ||
| BC086791.1 | - | 4 | 625 | |||
| D85036.1 | - | 4 | 626 | |||

Ak4
Genomic Map