siRNA Details
| Gene Symbol: CXCL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002089.3 | NP_002080.1 | 3 | 434 | ||
| GenBank mRNA | AF043340.1 | - | 2 | 172 | ||
| AK308504.1 | - | 2 | 523 | |||
| BC015753.1 | - | 3 | 338 | 4875789 | ||
| BX334969.2 | - | 3 | 444 | |||
| CR542171.1 | - | 3 | 274 | |||
| M36820.1 | - | 3 | 348 | |||
| M57731.1 | - | 3 | 343 | |||
| X53799.1 | - | 3 | 316 | |||

CXCL2
Genomic Map