siRNA Details
Gene Symbol: Abcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007435.1 | NP_031461.1 | 4 | 1743 | ||
GenBank mRNA | AK088792.1 | - | 4 | 1613 | ||
AK153589.1 | - | 4 | 1591 | |||
BC011273.1 | - | 4 | 1562 | 3710503 | ||
BC079840.1 | - | 4 | 1621 | 6810147 | ||
Z33637.1 | - | 4 | 1743 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
