siRNA Details
| Gene Symbol: Amelx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081978.2 | NP_001075447.1 | 5 | 402 | ||
| NM_001290371.1 | NP_001277300.1 | 5 | 330 | |||
| NM_009666.4 | NP_033796.1 | 5 | 402 | |||
| GenBank mRNA | AK029358.1 | - | 5 | 406 | ||
| BC059090.1 | - | 5 | 372 | 30286944 | ||
| CD773419.1 | - | 5 | 406 | |||
| D31768.1 | - | 5 | 393 | |||
| D31769.1 | - | 5 | 321 | |||
| KC731591.1 | - | 5 | 330 | |||
| M10095.1 | - | 2 | 210 | |||

Amelx
Genomic Map