siRNA Details
| Gene Symbol: Amh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007445.2 | NP_031471.2 | 3 | 580 | ||
| GenBank mRNA | AY911505.1 | - | 3 | 580 | ||
| BC150154.1 | - | 3 | 579 | |||
| BC150477.1 | - | 3 | 579 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amh| Gene Symbol: Amh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007445.2 | NP_031471.2 | 3 | 580 | ||
| GenBank mRNA | AY911505.1 | - | 3 | 580 | ||
| BC150154.1 | - | 3 | 579 | |||
| BC150477.1 | - | 3 | 579 | |||