siRNA Details
| Gene Symbol: Mat1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133653.3 | NP_598414.1 | 6 | 1007 | ||
| GenBank mRNA | AK149542.1 | - | 5 | 758 | ||
| AK153832.1 | - | 5 | 761 | |||
| BC011211.1 | - | 5 | 745 | 4196928 | ||
| L13622.1 | - | 5 | 759 | |||

Mat1a
Genomic Map