siRNA Details
| Gene Symbol: Ank3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009670.4 | NP_033800.2 | 18 | 3020 | ||
| NM_146005.3 | NP_666117.2 | 41 | 6079 | |||
| NM_170687.3 | NP_733788.2 | 18 | 3608 | |||
| NM_170688.2 | NP_733789.1 | 41 | 5491 | |||
| NM_170689.2 | NP_733790.2 | 40 | 6025 | |||
| NM_170690.2 | NP_733791.2 | 40 | 6016 | |||
| NM_170728.2 | NP_733924.2 | 39 | 5374 | |||
| NM_170729.2 | NP_733925.2 | 39 | 5962 | |||
| NM_170730.2 | NP_733926.2 | 16 | 2710 | |||
| GenBank mRNA | AF212924.1 | - | 16 | 2210 | ||
| AK033389.1 | - | 3 | 906 | |||
| AK035681.1 | - | 18 | 3336 | |||
| BC021657.1 | - | 39 | 5244 | 4188590 | ||
| BC026152.1 | - | 18 | 2622 | |||
| BC062122.1 | - | 7 | 3328 | |||
| L40631.1 | - | 18 | 3611 | |||
| L40632.1 | - | 41 | 5621 | |||
| U89274.1 | - | 20 | 2637 | |||
| U89275.1 | - | 20 | 3225 | |||

Ank3
Genomic Map