siRNA Details
| Gene Symbol: Ap1g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301211.1 | NP_001288140.1 | 8 | 1165 | ||
| NM_009677.6 | NP_033807.2 | 9 | 1174 | |||
| GenBank mRNA | AK034198.1 | - | 6 | 403 | ||
| AK036387.1 | - | 9 | 1117 | |||
| AK145821.1 | - | 8 | 1096 | |||
| BC006691.1 | - | 8 | 841 | |||
| BC052703.1 | - | 9 | 1047 | 5698075 | ||
| BC054535.1 | - | 9 | 1047 | 5698075 | ||
| X54424.1 | - | 8 | 1087 | |||

Ap1g1
Genomic Map