siRNA Details
| Gene Symbol: Ap2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077264.1 | NP_001070732.1 | 4 | 543 | ||
| NM_007458.2 | NP_031484.1 | 4 | 543 | |||
| GenBank mRNA | AK135424.1 | - | 4 | 472 | ||
| AK155515.1 | - | 4 | 479 | |||
| BC031433.1 | - | 4 | 493 | 4481643 | ||
| CA327188.1 | - | 4 | 549 | |||

Ap2a1
Genomic Map