siRNA Details
| Gene Symbol: Ap3s2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009682.3 | NP_033812.3 | 5 | 451 | ||
| GenBank mRNA | AK028372.1 | - | 5 | 434 | ||
| AK083911.1 | - | 5 | 440 | |||
| AK150085.1 | - | 5 | 444 | |||
| AK214389.1 | - | 3 | 193 | |||
| BC060236.1 | - | 5 | 412 | 6853219 | ||
| U91933.1 | - | 5 | 406 | |||

Ap3s2
Genomic Map