siRNA Details
| Gene Symbol: Ap4s1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021710.3 | NP_068356.1 | 6 | 749 | ||
| GenBank mRNA | AF092093.1 | - | 6 | 678 | ||
| AK005283.1 | - | 6 | 499 | |||
| AK178071.1 | - | 5 | 297 | |||
| BC053339.1 | - | 6 | 509 | 5370721 | ||
| BP426557.1 | - | 4 | 204 | |||

Ap4s1
Genomic Map