siRNA Details
| Gene Symbol: Apaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042558.1 | NP_001036023.1 | 20 | 3480 | ||
| NM_001282947.1 | NP_001269876.1 | 20 | 3404 | |||
| NM_009684.2 | NP_033814.2 | 20 | 3437 | |||
| GenBank mRNA | AF064071.1 | - | 20 | 3364 | ||
| AK220340.1 | - | 20 | 3476 | |||
| BC083191.1 | - | 20 | 3355 | |||
| BC131683.1 | - | 20 | 2996 | 40111198 | ||
| BC131684.1 | - | 20 | 2996 | 40111199 | ||

Apaf1
Genomic Map