siRNA Details
| Gene Symbol: Apaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001042558.1 | NP_001036023.1 | 27 | 4368 | ||
| NM_001282947.1 | NP_001269876.1 | 27 | 4292 | |||
| NM_009684.2 | NP_033814.2 | 27 | 4325 | |||
| GenBank mRNA | AF064071.1 | - | 27 | 4252 | ||
| AK220340.1 | - | 27 | 4364 | |||
| BC083191.1 | - | 27 | 4243 | |||
| BC131683.1 | - | 27 | 3884 | 40111198 | ||
| BC131684.1 | - | 27 | 3884 | 40111199 | ||
| DT924066.1 | - | 2 | 151 | |||

Apaf1
Genomic Map