siRNA Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253885.1 | NP_001240814.1 | 7 | 1591 | ||
| NM_001253886.1 | NP_001240815.1 | 7 | 1591 | |||
| NM_001253887.1 | NP_001240816.1 | 7 | 591 | |||
| NM_009685.3 | NP_033815.1 | 7 | 1405 | |||
| GenBank mRNA | AF206720.1 | - | 7 | 1314 | ||
| AK030748.1 | - | 7 | 1288 | |||
| AK075730.1 | - | 7 | 571 | |||
| AK083830.1 | - | 7 | 1321 | |||
| AK164140.1 | - | 7 | 1316 | |||
| BC036956.1 | - | 6 | 1229 | |||
| BC048395.1 | - | 5 | 737 | 6392016 | ||

Apbb1
Genomic Map