siRNA Details
| Gene Symbol: Apbb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253885.1 | NP_001240814.1 | 2 | 780 | ||
| NM_001253886.1 | NP_001240815.1 | 2 | 780 | |||
| NM_009685.3 | NP_033815.1 | 2 | 594 | |||
| GenBank mRNA | AF206720.1 | - | 2 | 503 | ||
| AK030748.1 | - | 2 | 477 | |||
| AK083830.1 | - | 2 | 510 | |||
| AK144340.1 | - | 2 | 782 | |||
| AK164140.1 | - | 2 | 505 | |||
| BC036956.1 | - | 1 | 418 | |||

Apbb1
Genomic Map