siRNA Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201413.1 | NP_001188342.1 | 13 | 2276 | ||
| NM_001201414.1 | NP_001188343.1 | 12 | 2279 | |||
| NM_001201415.1 | NP_001188344.1 | 12 | 2213 | |||
| NM_001201416.1 | NP_001188345.1 | 3 | 379 | |||
| NM_009686.2 | NP_033816.1 | 14 | 2282 | |||
| GenBank mRNA | AK004792.1 | - | 14 | 2136 | ||
| AK153260.1 | - | 12 | 2245 | |||
| AK167649.1 | - | 13 | 2141 | |||
| AK169959.1 | - | 12 | 2154 | |||
| AK172405.1 | - | 3 | 383 | |||
| BC042797.1 | - | 4 | 421 | |||
| BC076587.1 | - | 12 | 2059 | 30550087 | ||

Apbb2
Genomic Map