siRNA Details
| Gene Symbol: Xiap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001301639.1 | NP_001288568.1 | 2 | 1316 | ||
| NM_001301641.1 | NP_001288570.1 | 2 | 1103 | |||
| NM_009688.3 | NP_033818.2 | 3 | 1188 | |||
| GenBank mRNA | BC138528.1 | - | 1 | 1111 | 8861548 | |
| BC145861.1 | - | 1 | 1111 | 40131164 | ||
| U36842.1 | - | 2 | 1020 | |||
| U88990.1 | - | 1 | 1480 | |||

Xiap
Genomic Map