siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079865.1 | - | 4 | 437 | ||
| AK134480.1 | - | 4 | 490 | |||
| AK137466.1 | - | 4 | 519 | |||
| AK153934.1 | - | 4 | 463 | |||
| AK160874.1 | - | 4 | 487 | |||
| AK163586.1 | - | 4 | 494 | |||
| AK167315.1 | - | 4 | 463 | |||
| AK167375.1 | - | 4 | 503 | |||
| BC007133.1 | - | 4 | 482 | 3594358 | ||
| U35846.1 | - | 4 | 447 | |||

Api5
Genomic Map