siRNA Details
| Gene Symbol: Api5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK079865.1 | - | 7 | 877 | ||
| AK134480.1 | - | 7 | 929 | |||
| AK137466.1 | - | 7 | 958 | |||
| AK153934.1 | - | 7 | 902 | |||
| AK160874.1 | - | 7 | 926 | |||
| AK163586.1 | - | 6 | 1360 | |||
| AK167315.1 | - | 7 | 902 | |||
| AK167375.1 | - | 7 | 942 | |||
| AK210073.1 | - | 1 | 23 | |||
| BC007133.1 | - | 7 | 921 | 3594358 | ||
| U35846.1 | - | 7 | 886 | |||

Api5
Genomic Map