siRNA Details
Gene Symbol: Apoc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110009.2 | NP_001103479.1 | 3 | 259 | ||
NM_007469.5 | NP_031495.1 | 2 | 381 | |||
GenBank mRNA | AK011358.1 | - | 2 | 296 | ||
BC019398.1 | - | 3 | 234 | 5124986 | ||
BC035445.1 | - | 3 | 246 | |||
BC094638.1 | - | 2 | 228 | 875610 | ||
BY087738.1 | - | 3 | 260 | |||
CB947285.1 | - | 2 | 348 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
