siRNA Details
| Gene Symbol: Apoc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289755.1 | NP_001276684.1 | 4 | 359 | ||
| NM_001289756.1 | NP_001276685.1 | 4 | 329 | |||
| NM_001289833.1 | NP_001276762.1 | 3 | 187 | |||
| NM_023114.4 | NP_075603.1 | 4 | 255 | |||
| GenBank mRNA | AK002908.1 | - | 4 | 254 | ||
| AK008260.1 | - | 4 | 258 | |||

Apoc3
Genomic Map