siRNA Details
| Gene Symbol: Aqp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009701.4 | NP_033831.1 | 1 | 375 | ||
| GenBank mRNA | AF087654.1 | - | 1 | 624 | ||
| AK004736.1 | - | 1 | 258 | |||
| AK009302.1 | - | 1 | 260 | |||
| BC150769.1 | - | 2 | 421 | 9087680 | ||

Aqp5
Genomic Map