siRNA Details
| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 1800 | |||
| M37890.1 | - | 3 | 1739 | |||
| S56585.1 | - | 3 | 1818 | |||
| X53779.1 | - | 3 | 1771 | |||
| X59411.1 | - | 3 | 1770 | |||
| X59592.1 | - | 3 | 1859 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ar| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 1800 | |||
| M37890.1 | - | 3 | 1739 | |||
| S56585.1 | - | 3 | 1818 | |||
| X53779.1 | - | 3 | 1771 | |||
| X59411.1 | - | 3 | 1770 | |||
| X59592.1 | - | 3 | 1859 | |||