siRNA Details
| Gene Symbol: Arc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276684.1 | NP_001263613.1 | 1 | 277 | ||
| NM_018790.3 | NP_061260.1 | 1 | 277 | |||
| GenBank mRNA | AF162777.1 | - | 1 | 268 | ||
| AK157822.1 | - | 1 | 278 | |||
| AK170446.1 | - | 1 | 277 | |||
| BC023127.1 | - | 1 | 266 | 3498057 | ||

Arc
Genomic Map