siRNA Details
| Gene Symbol: Arf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031259.1 | - | 4 | 389 | ||
| AK042977.1 | - | 4 | 429 | |||
| AK053123.1 | - | 4 | 847 | |||
| AK077591.1 | - | 4 | 402 | |||
| AK167133.1 | - | 4 | 399 | |||
| AK167601.1 | - | 4 | 403 | |||
| AK167653.1 | - | 4 | 402 | |||
| AK169094.1 | - | 4 | 399 | |||
| AK169412.1 | - | 4 | 399 | |||
| BC010487.1 | - | 4 | 682 | 3483748 | ||
| D87899.1 | - | 4 | 394 | |||

Arf2
Genomic Map