siRNA Details
| Gene Symbol: Arf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007481.3 | NP_031507.1 | 2 | 695 | ||
| GenBank mRNA | AK006366.1 | - | 1 | 406 | ||
| AK156960.1 | - | 2 | 680 | |||
| BC003478.1 | - | 2 | 657 | 2655889 | ||
| BC083112.1 | - | 2 | 695 | 5354797 | ||
| D87903.1 | - | 2 | 637 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_024152, BC091146, L12385.

Arf6
Genomic Map